Learn More
Grainger Externalooving Toolholder KGDL 1616H
Catalog No. 502764381
Encompass_Preferred
The Kyocera External Grooving Toolholder 61NR09 is a right-hand toolholder for external grooving and turning. It has a shank diameter of 1" and an overall length of 5". The toolholder accepts inserts with a thickness of 3/64" to 5/64" and a nose radius of 1/32" or 1/64". It is made of steel and has a black oxide finish. The toolholder allows high precision grooving, profiling, and turning operations. It has a maximum cutting depth of 0.630" and can machine a variety of materials including steel, stainless steel, cast iron, and non-ferrous metals. The toolholder features a screw clamping system for secure insert retention.
Catalog No. 50-276-4381 Supplier Grainger Supplier No. 170JK9
The Kyocera External Grooving Toolholder 61NR09 is a right-hand toolholder for external grooving and turning. It has a shank diameter of 1" and an overall length of 5". The toolholder accepts inserts with a thickness of 3/64" to 5/64" and a nose radius of 1/32" or 1/64". It is made of steel and has a black oxide finish. The toolholder allows high precision grooving, profiling, and turning operations. It has a maximum cutting depth of 0.630" and can machine a variety of materials including steel, stainless steel, cast iron, and non-ferrous metals. The toolholder features a screw clamping system for secure insert retention.