Learn More
Grainger Externalooving Toolholder KGDR 2020K
Catalog No. 502764657
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JM3 is a toolholder designed for external grooving operations. It has a JM3 style toolholder interface and accepts inserts with a .118" thickness. The toolholder body is made from steel and has an overall length of 3.5". It is compatible with a variety of Kyocera grooving inserts including the DGN300 series. The toolholder has a maximum depth of cut capability of 0.197" and can be run at speeds up to 1965 SFM. It is an indexable toolholder solution for external grooving applications.
Catalog No. 50-276-4657 Supplier Grainger Supplier No. 170JM3
The Kyocera External Grooving Toolholder 170JM3 is a toolholder designed for external grooving operations. It has a JM3 style toolholder interface and accepts inserts with a .118" thickness. The toolholder body is made from steel and has an overall length of 3.5". It is compatible with a variety of Kyocera grooving inserts including the DGN300 series. The toolholder has a maximum depth of cut capability of 0.197" and can be run at speeds up to 1965 SFM. It is an indexable toolholder solution for external grooving applications.