Learn More
Grainger Externalooving Toolholder KGDR 16X3T
Catalog No. 502764658
Encompass_Preferred
The 170JR2 is a Dayton 1/3 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Sleeve Bearing Electric Motor. It has a 1/3 horsepower, operates on 115 volts at 60 Hz, has a 1725 RPM at full load, and a 56C frame size. The motor is open drip proof with class B insulation and ball bearings on the drive end and sleeve bearings on the opposite end. It can be used for general purpose applications.
Catalog No. 50-276-4658 Supplier Grainger Supplier No. 170JR2
The 170JR2 is a Dayton 1/3 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Sleeve Bearing Electric Motor. It has a 1/3 horsepower, operates on 115 volts at 60 Hz, has a 1725 RPM at full load, and a 56C frame size. The motor is open drip proof with class B insulation and ball bearings on the drive end and sleeve bearings on the opposite end. It can be used for general purpose applications.