Learn More
Grainger Externalooving Toolholder KGDR 1616H
Catalog No. 502764192
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JL9 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and accepts grooving inserts with thicknesses ranging from 0.078 to 0.197 inches. It is made from steel and has an ISO designation of PCLNR/L 2020K12. The toolholder allows both right hand and left hand grooving.
Catalog No. 50-276-4192 Supplier Grainger Supplier No. 170JL9
The Kyocera External Grooving Toolholder 170JL9 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and accepts grooving inserts with thicknesses ranging from 0.078 to 0.197 inches. It is made from steel and has an ISO designation of PCLNR/L 2020K12. The toolholder allows both right hand and left hand grooving.