Learn More
Grainger Externalooving Toolholder KGDR 2020C
Catalog No. 502764302
Encompass_Preferred
The Kyocera Indexable Parting and Grooving Insert is a carbide insert designed for parting and grooving operations. It features a TiAlN PVD coating for wear resistance and heat resistance. The insert has a thickness of 0.118" and is available in widths ranging from 0.039" to 0.315". It has an ISO shape code of TT and is compatible with tool holders that accept TT inserts. The part number for this insert is 170JM2. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The insert provides accurate grooving and excellent surface finishes.
Catalog No. 50-276-4302 Supplier Grainger Supplier No. 170JM2
The Kyocera Indexable Parting and Grooving Insert is a carbide insert designed for parting and grooving operations. It features a TiAlN PVD coating for wear resistance and heat resistance. The insert has a thickness of 0.118" and is available in widths ranging from 0.039" to 0.315". It has an ISO shape code of TT and is compatible with tool holders that accept TT inserts. The part number for this insert is 170JM2. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The insert provides accurate grooving and excellent surface finishes.