Learn More
Grainger Externalooving Toolholder KGDR 123T2
Catalog No. 502764172
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JJ0 is a toolholder designed for external grooving operations. It features a JIS B 6339-3 MDJNR 2020K-16 clamping system and accepts Kyocera CNMG style inserts with a thickness of 2.5mm. The toolholder has an overall length of 4 inches and a shank diameter of 1 inch. It is made from forged steel for durability and rigidity. The 170JJ0 is suitable for grooving, parting and cutoff applications in a variety of materials. It can provide excellent chip control when paired with the proper insert geometry and grades. This toolholder enables secure clamping of inserts for accurate grooving.
Catalog No. 50-276-4172 Supplier Grainger Supplier No. 170JJ0
The Kyocera External Grooving Toolholder 170JJ0 is a toolholder designed for external grooving operations. It features a JIS B 6339-3 MDJNR 2020K-16 clamping system and accepts Kyocera CNMG style inserts with a thickness of 2.5mm. The toolholder has an overall length of 4 inches and a shank diameter of 1 inch. It is made from forged steel for durability and rigidity. The 170JJ0 is suitable for grooving, parting and cutoff applications in a variety of materials. It can provide excellent chip control when paired with the proper insert geometry and grades. This toolholder enables secure clamping of inserts for accurate grooving.