Learn More
Grainger Externalooving Toolholder KGDR 2525M
Catalog No. 502764343
Encompass_Preferred
The Kyocera External Grooving Toolholder 61NR08 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a nominal groove width capacity of 0.078-0.315 inches and a grooving depth capacity of 0.039-0.197 inches. It is made of steel and is uncoated. The toolholder accepts ISO turning inserts with a thickness of 0.078-0.197 inches. It can provide rigid clamping of inserts for interrupted cuts. The 61NR08 is ideal for external grooving, cutoff and parting applications.
Catalog No. 50-276-4343 Supplier Grainger Supplier No. 170JN7
The Kyocera External Grooving Toolholder 61NR08 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a nominal groove width capacity of 0.078-0.315 inches and a grooving depth capacity of 0.039-0.197 inches. It is made of steel and is uncoated. The toolholder accepts ISO turning inserts with a thickness of 0.078-0.197 inches. It can provide rigid clamping of inserts for interrupted cuts. The 61NR08 is ideal for external grooving, cutoff and parting applications.