Learn More
Grainger Externalooving Toolholder KGDR 2525M
Catalog No. 502764421
Encompass_Preferred
The Kyocera External Grooving Toolholder is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and can hold grooving inserts with thicknesses between 0.040-0.125 inches. It is made of steel and has an American National Standard Code (ANSI) style insert seat. The toolholder allows for high precision grooving in a variety of materials.
Catalog No. 50-276-4421 Supplier Grainger Supplier No. 170JN6
The Kyocera External Grooving Toolholder is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder has a maximum grooving depth capacity of 0.315 inches and can hold grooving inserts with thicknesses between 0.040-0.125 inches. It is made of steel and has an American National Standard Code (ANSI) style insert seat. The toolholder allows for high precision grooving in a variety of materials.