Learn More
Grainger Externalooving Toolholder KGDR 166T3
Catalog No. 502764437
Encompass_Preferred
The 170JK6 is a 6-inch diameter, 3-blade, right-hand rotation propeller fan manufactured by Aerovent. It features a cast aluminum alloy construction with an enclosed airfoil blade design and is balanced in accordance with AMCA Standard 204-05. This direct drive fan has a maximum speed of 3500 RPM and is designed for panel mounting via the integral flange. With an airflow capacity of 850 CFM and 0.25 inches static pressure, this fan is suitable for industrial ventilation and cooling applications. The maximum temperature rating is 250°F.
Catalog No. 50-276-4437 Supplier Grainger Supplier No. 170JK6
The 170JK6 is a 6-inch diameter, 3-blade, right-hand rotation propeller fan manufactured by Aerovent. It features a cast aluminum alloy construction with an enclosed airfoil blade design and is balanced in accordance with AMCA Standard 204-05. This direct drive fan has a maximum speed of 3500 RPM and is designed for panel mounting via the integral flange. With an airflow capacity of 850 CFM and 0.25 inches static pressure, this fan is suitable for industrial ventilation and cooling applications. The maximum temperature rating is 250°F.