Learn More
Grainger Externalooving Toolholder KGDR 2020K
Catalog No. 502764440
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JM7 is a high precision toolholder for external grooving and parting. It has a JM7 interface and is compatible with ISO turning holders. The toolholder body is made from forged steel for durability. It accepts inserts with a thickness of 2.5mm and widths up to 3.97mm. The maximum depth of cut is 1mm. This is a high quality toolholder suitable for precise grooving and parting operations.
Catalog No. 50-276-4440 Supplier Grainger Supplier No. 170JM7
The Kyocera External Grooving Toolholder 170JM7 is a high precision toolholder for external grooving and parting. It has a JM7 interface and is compatible with ISO turning holders. The toolholder body is made from forged steel for durability. It accepts inserts with a thickness of 2.5mm and widths up to 3.97mm. The maximum depth of cut is 1mm. This is a high quality toolholder suitable for precise grooving and parting operations.