Learn More
Grainger Externalooving Toolholder KGDR 103JX
Catalog No. 502763816
Encompass_Preferred
The KYOCERA Indexable Parting and Grooving Insert is a high-performance parting and grooving insert for use on steel, stainless steel, cast iron, and non-ferrous metals. It features a sharp edge and chipbreaker for smooth parting and grooving operations. The insert is made of micro-grain carbide with a TiAlN coating for wear resistance and heat resistance. It has a thickness of 2.5mm and is available in widths from 2.5mm to 4mm for creating grooves and slots. The insert shape allows bidirectional machining. Overall, this is a durable and versatile parting and grooving insert suitable for demanding applications on various materials.
Catalog No. 50-276-3816 Supplier Grainger Supplier No. 170JW3
The KYOCERA Indexable Parting and Grooving Insert is a high-performance parting and grooving insert for use on steel, stainless steel, cast iron, and non-ferrous metals. It features a sharp edge and chipbreaker for smooth parting and grooving operations. The insert is made of micro-grain carbide with a TiAlN coating for wear resistance and heat resistance. It has a thickness of 2.5mm and is available in widths from 2.5mm to 4mm for creating grooves and slots. The insert shape allows bidirectional machining. Overall, this is a durable and versatile parting and grooving insert suitable for demanding applications on various materials.