Learn More
Grainger Externalooving Toolholder KGDR 1212J
Catalog No. 502763817
Encompass_Preferred
The Kyocera External Grooving Toolholder is a toolholder designed for external grooving operations. It features a square shank design and accepts grooving inserts with a thickness of 2.5mm. The toolholder is made from steel and has an overall length of 4 inches. It is compatible with ISO turning holders and features a lever lock design for quick insert changes. The maximum depth of groove is 0.472 inches and it has a maximum grooving width capacity of 0.315 inches. This is a high quality toolholder suitable for precise grooving applications.
Catalog No. 50-276-3817 Supplier Grainger Supplier No. 170JW7
The Kyocera External Grooving Toolholder is a toolholder designed for external grooving operations. It features a square shank design and accepts grooving inserts with a thickness of 2.5mm. The toolholder is made from steel and has an overall length of 4 inches. It is compatible with ISO turning holders and features a lever lock design for quick insert changes. The maximum depth of groove is 0.472 inches and it has a maximum grooving width capacity of 0.315 inches. This is a high quality toolholder suitable for precise grooving applications.