Learn More
Grainger Externalooving Toolholder KGDR 3232X
Catalog No. 502764244
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JU7 is a toolholder designed for external grooving operations. It has a JIS standard tool shank type with a designation of JETECLCR/L 1707 and accepts grooving tool bits with a width up to 2mm. The toolholder body is made from steel for durability. It has a maximum grooving depth capability of 4mm and can be used at speeds up to 15,000 RPM. The overall toolholder length is 170mm. This is a high precision toolholder well-suited for demanding grooving applications.
Catalog No. 50-276-4244 Supplier Grainger Supplier No. 170JU7
The Kyocera External Grooving Toolholder 170JU7 is a toolholder designed for external grooving operations. It has a JIS standard tool shank type with a designation of JETECLCR/L 1707 and accepts grooving tool bits with a width up to 2mm. The toolholder body is made from steel for durability. It has a maximum grooving depth capability of 4mm and can be used at speeds up to 15,000 RPM. The overall toolholder length is 170mm. This is a high precision toolholder well-suited for demanding grooving applications.