Learn More
Grainger Externalooving Toolholder KGDR 1616H
Catalog No. 502764271
Encompass_Preferred
The KYOCERA External Grooving Toolholder 170JM1 is a toolholder designed for external grooving operations. It has a JM style toolholder interface and accepts inserts with a .118" thickness. The toolholder body is made from steel and has an overall length of 3.5". It is compatible with KYOCERA's JM style grooving inserts that have a .118" thickness. The toolholder has an optimal cutting depth range of .039" to .197" and can be used for grooving, parting and cutoff applications. It is suitable for use on most lathes and can hold inserts securely for accurate grooving and parting work.
Catalog No. 50-276-4271 Supplier Grainger Supplier No. 170JM0
The KYOCERA External Grooving Toolholder 170JM1 is a toolholder designed for external grooving operations. It has a JM style toolholder interface and accepts inserts with a .118" thickness. The toolholder body is made from steel and has an overall length of 3.5". It is compatible with KYOCERA's JM style grooving inserts that have a .118" thickness. The toolholder has an optimal cutting depth range of .039" to .197" and can be used for grooving, parting and cutoff applications. It is suitable for use on most lathes and can hold inserts securely for accurate grooving and parting work.