Learn More
Grainger Externalooving Toolholder KGDR 12X3T
Catalog No. 502764177
Encompass_Preferred
The 170JT2 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof Fan Motor manufactured by Dayton Electric Mfg. Co. It has a 1/3 horsepower, operates on 115/230 volt AC power, has a speed of 1725 RPM, uses a 56C frame, and has an open drip proof enclosure. The motor is suitable for exhaust, cooling, ventilation and material handling applications.
Catalog No. 50-276-4177 Supplier Grainger Supplier No. 170JT2
The 170JT2 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof Fan Motor manufactured by Dayton Electric Mfg. Co. It has a 1/3 horsepower, operates on 115/230 volt AC power, has a speed of 1725 RPM, uses a 56C frame, and has an open drip proof enclosure. The motor is suitable for exhaust, cooling, ventilation and material handling applications.