Learn More
Grainger Externalooving Toolholder KGDR 1212F
Catalog No. 502764239
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JX1 is a toolholder for external grooving operations. It has a shank diameter of 1 inch, an overall length of 5 inches, and accepts inserts with a thickness of 0.118 inches. The toolholder is made of steel and has a black oxide finish. It can be used for a variety of materials including steel, stainless steel, cast iron, and non-ferrous metals. The 170JX1 provides rigid clamping of inserts for accurate grooving, threading, and cutoff operations. This versatile toolholder is ideal for use on CNC lathes, screw machines, and manual lathes.
Catalog No. 50-276-4239 Supplier Grainger Supplier No. 170JX1
The Kyocera External Grooving Toolholder 170JX1 is a toolholder for external grooving operations. It has a shank diameter of 1 inch, an overall length of 5 inches, and accepts inserts with a thickness of 0.118 inches. The toolholder is made of steel and has a black oxide finish. It can be used for a variety of materials including steel, stainless steel, cast iron, and non-ferrous metals. The 170JX1 provides rigid clamping of inserts for accurate grooving, threading, and cutoff operations. This versatile toolholder is ideal for use on CNC lathes, screw machines, and manual lathes.