Learn More
Grainger Externalooving Toolholder KGDR 1212F
Catalog No. 502764267
Encompass_Preferred
The Kyocera External Grooving Toolholder 170JW6 is a toolholder designed for external grooving operations. It has a JIS standard tool shank type W with a nominal shank width of 20mm. The toolholder body is made from steel and allows a maximum grooving depth of 4mm. It accepts ISO standard triangular insert shapes with a thickness of 2.5mm and has a maximum insert width capacity of 16mm. The toolholder enables high precision grooving, parting and cutoff operations on CNC machine tools. Its precision ground body provides stability and rigidity for smooth machining.
Catalog No. 50-276-4267 Supplier Grainger Supplier No. 170JW6
The Kyocera External Grooving Toolholder 170JW6 is a toolholder designed for external grooving operations. It has a JIS standard tool shank type W with a nominal shank width of 20mm. The toolholder body is made from steel and allows a maximum grooving depth of 4mm. It accepts ISO standard triangular insert shapes with a thickness of 2.5mm and has a maximum insert width capacity of 16mm. The toolholder enables high precision grooving, parting and cutoff operations on CNC machine tools. Its precision ground body provides stability and rigidity for smooth machining.