Learn More
Grainger Externalooving Toolholder KGDR 2525M
Catalog No. 502764382
Encompass_Preferred
The Kyocera External Grooving Toolholder 61NR04 is a toolholder designed for external grooving operations. It has a shank diameter of 1 inch, an overall length of 4 inches, and accepts inserts with a thickness of 0.118 inches. The toolholder is made of steel and has a black oxide finish. It can be used for turning, facing, profiling and external grooving applications. The toolholder has a screw clamping system for securing inserts. It is compatible with Kyocera's TNGA160 series of inserts for grooving.
Catalog No. 50-276-4382 Supplier Grainger Supplier No. 170JN0
The Kyocera External Grooving Toolholder 61NR04 is a toolholder designed for external grooving operations. It has a shank diameter of 1 inch, an overall length of 4 inches, and accepts inserts with a thickness of 0.118 inches. The toolholder is made of steel and has a black oxide finish. It can be used for turning, facing, profiling and external grooving applications. The toolholder has a screw clamping system for securing inserts. It is compatible with Kyocera's TNGA160 series of inserts for grooving.