Learn More
Grainger Externalooving Toolholder KKCL 163C
Catalog No. 502764755
Encompass_Preferred
The KYOCERA External Grooving Toolholder KKCL-163C-170HX7 is a high-quality toolholder designed for external grooving operations. It features a KKCL style toolholder with a clamping diameter of 0.63 inches and an overall length of 6.7 inches. The toolholder has an IC insert pocket that accepts IC908 style inserts with a thickness of 0.078 inches. It is made from precision-ground steel for rigidity and accuracy. The KKCL-163C-170HX7 has an optimal cutting depth of 0.039 inches and can machine a groove width range of 0.039-0.197 inches. This durable toolholder allows for high-precision grooving in a variety of materials.
Catalog No. 50-276-4755 Supplier Grainger Supplier No. 170HX7
The KYOCERA External Grooving Toolholder KKCL-163C-170HX7 is a high-quality toolholder designed for external grooving operations. It features a KKCL style toolholder with a clamping diameter of 0.63 inches and an overall length of 6.7 inches. The toolholder has an IC insert pocket that accepts IC908 style inserts with a thickness of 0.078 inches. It is made from precision-ground steel for rigidity and accuracy. The KKCL-163C-170HX7 has an optimal cutting depth of 0.039 inches and can machine a groove width range of 0.039-0.197 inches. This durable toolholder allows for high-precision grooving in a variety of materials.