Learn More
Grainger Externalooving Toolholder KKCL 62CF
Catalog No. 502763837
Encompass_Preferred
The Kyocera External Grooving Toolholder 170HY2 is a toolholder for external grooving and turning applications. It has a 170HY2 insert style and is made of steel with a black oxide finish. The toolholder has a 1/2 inch shank diameter and accepts inserts with a thickness of 0.118 inches. It has a maximum grooving depth of 0.315 inches and a maximum grooving width of 0.197 inches. The toolholder allows high precision grooving and turning in a wide range of materials.
Catalog No. 50-276-3837 Supplier Grainger Supplier No. 170HY2
The Kyocera External Grooving Toolholder 170HY2 is a toolholder for external grooving and turning applications. It has a 170HY2 insert style and is made of steel with a black oxide finish. The toolholder has a 1/2 inch shank diameter and accepts inserts with a thickness of 0.118 inches. It has a maximum grooving depth of 0.315 inches and a maximum grooving width of 0.197 inches. The toolholder allows high precision grooving and turning in a wide range of materials.