Learn More
Grainger Externalooving Toolholder KKCL 162D
Catalog No. 502764352
Encompass_Preferred
The KYOCERA Internal Grooving Toolholder 170HX6 is a toolholder designed for internal grooving operations. It has a maximum grooving depth of 0.787" and a minimum grooving depth of 0.039". The toolholder has an American National Standards Institute (ANSI) style shank with a shank height of 1.260" and shank width of 1.260". It is made of steel and accepts inserts with a thickness of 0.118". The toolholder allows high precision internal grooving in a variety of materials.
Catalog No. 50-276-4352 Supplier Grainger Supplier No. 170HX6
The KYOCERA Internal Grooving Toolholder 170HX6 is a toolholder designed for internal grooving operations. It has a maximum grooving depth of 0.787" and a minimum grooving depth of 0.039". The toolholder has an American National Standards Institute (ANSI) style shank with a shank height of 1.260" and shank width of 1.260". It is made of steel and accepts inserts with a thickness of 0.118". The toolholder allows high precision internal grooving in a variety of materials.