Learn More
Grainger Externalooving Toolholder KKCL 163D
Catalog No. 502764353
Encompass_Preferred
The Kyocera External Grooving Toolholder 170HX8 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder accepts inserts with a thickness of 0.118 inches and features a screw clamp to securely hold inserts in place. It is made of steel and can be used for high speed steel or carbide inserts. The 170HX8 has an American National Standards Institute (ANSI) standard toolholder body for versatility and compatibility with tooling systems. This external grooving toolholder enables precise grooving, parting and cutoff operations.
Catalog No. 50-276-4353 Supplier Grainger Supplier No. 170HX8
The Kyocera External Grooving Toolholder 170HX8 is a toolholder designed for external grooving operations. It has a shank diameter of 0.625 inches and an overall length of 4 inches. The toolholder accepts inserts with a thickness of 0.118 inches and features a screw clamp to securely hold inserts in place. It is made of steel and can be used for high speed steel or carbide inserts. The 170HX8 has an American National Standards Institute (ANSI) standard toolholder body for versatility and compatibility with tooling systems. This external grooving toolholder enables precise grooving, parting and cutoff operations.