Learn More
Grainger Externalooving Toolholder KKCR 162C
Catalog No. 502763841
Encompass_Preferred
The Grainger 170HZ2 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Electric Motor. It has a 1/3 horsepower, operates on 115/230 volt AC power, has a 1725 RPM speed, uses a 56C frame, is open drip proof, utilizes ball bearings, and is designed for general purpose applications. The motor is thermally protected, reversible, and suitable for OEM and MRO applications.
Catalog No. 50-276-3841 Supplier Grainger Supplier No. 170HZ2
Add to Cart
Add to Cart
The Grainger 170HZ2 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Electric Motor. It has a 1/3 horsepower, operates on 115/230 volt AC power, has a 1725 RPM speed, uses a 56C frame, is open drip proof, utilizes ball bearings, and is designed for general purpose applications. The motor is thermally protected, reversible, and suitable for OEM and MRO applications.