Learn More
Grainger Externalooving Toolholder KKCR 122B
Catalog No. 502763838
Encompass_Preferred
The External Grooving Toolholder KKCR-122B-170HY8 by Kennametal is a right hand external grooving tool holder with a 1/2" shank diameter and accepts inserts with a .122" thickness. It is made from Kennametal BeyondTM grade powdered metal which provides rigidity and heat resistance. The toolholder has a maximum depth of cut of 0.315" and can be used for external grooving, profiling and cutoff applications in steel, stainless steel, and cast iron. It has an overall length of 5.5" and weighs 0.44 lbs. The toolholder requires a screw clamp to hold the insert in place.
Catalog No. 50-276-3838 Supplier Grainger Supplier No. 170HY8
The External Grooving Toolholder KKCR-122B-170HY8 by Kennametal is a right hand external grooving tool holder with a 1/2" shank diameter and accepts inserts with a .122" thickness. It is made from Kennametal BeyondTM grade powdered metal which provides rigidity and heat resistance. The toolholder has a maximum depth of cut of 0.315" and can be used for external grooving, profiling and cutoff applications in steel, stainless steel, and cast iron. It has an overall length of 5.5" and weighs 0.44 lbs. The toolholder requires a screw clamp to hold the insert in place.