Learn More
Grainger Externalooving Toolholder KKCR 82DF
Catalog No. 502764419
Encompass_Preferred
The KYOCERA Internal Grooving Toolholder 170UK9 is a toolholder designed for internal grooving operations. It has a shank diameter of 25mm and an insert size of 9mm. The toolholder is made of steel and accepts triangular inserts with a thickness of 2.5mm. It has a maximum grooving depth of 4mm and a minimum grooving diameter of 17mm. The toolholder enables high-precision internal grooving in materials like steel, stainless steel, and cast iron. It is suitable for use on CNC lathes.
Catalog No. 50-276-4419 Supplier Grainger Supplier No. 170JA1
The KYOCERA Internal Grooving Toolholder 170UK9 is a toolholder designed for internal grooving operations. It has a shank diameter of 25mm and an insert size of 9mm. The toolholder is made of steel and accepts triangular inserts with a thickness of 2.5mm. It has a maximum grooving depth of 4mm and a minimum grooving diameter of 17mm. The toolholder enables high-precision internal grooving in materials like steel, stainless steel, and cast iron. It is suitable for use on CNC lathes.