Learn More
Grainger Externalooving Toolholder KKCR 163D
Catalog No. 502764800
Encompass_Preferred
The KYOCERA External Grooving Toolholder KKCR-163D-170HZ5 is a toolholder for external grooving and turning. It has a shank diameter of 0.63 inches, an overall length of 4.5 inches, and a grooving depth of 0.24 inches. The toolholder is made of steel and accepts inserts with a thickness of 0.08 inches. It has a positive rake angle of 11 degrees. The KKCR-163D-170HZ5 is suitable for both right-hand and left-hand turning operations.
Catalog No. 50-276-4800 Supplier Grainger Supplier No. 170HZ5
The KYOCERA External Grooving Toolholder KKCR-163D-170HZ5 is a toolholder for external grooving and turning. It has a shank diameter of 0.63 inches, an overall length of 4.5 inches, and a grooving depth of 0.24 inches. The toolholder is made of steel and accepts inserts with a thickness of 0.08 inches. It has a positive rake angle of 11 degrees. The KKCR-163D-170HZ5 is suitable for both right-hand and left-hand turning operations.