Learn More
Grainger Externalooving Toolholder KKCR 102DF
Catalog No. 502764278
Encompass_Preferred
The 170HY6 is a high-quality hydraulic oil filter made by Baldwin Filters. This spin-on filter has a collapse pressure of 25 PSI minimum, an operating temperature range of -20 to 250 degrees Fahrenheit, and a nominal micron rating of 25. It is designed to work with petroleum-based hydraulic fluids and has a height of 5.81 inches, an outside diameter of 3.74 inches, and 1-16 UNF threads. The 170HY6 filter can capture contaminants and prevent wear to help extend the life of hydraulic components.
Catalog No. 50-276-4278 Supplier Grainger Supplier No. 170HY6
Add to Cart
Add to Cart
The 170HY6 is a high-quality hydraulic oil filter made by Baldwin Filters. This spin-on filter has a collapse pressure of 25 PSI minimum, an operating temperature range of -20 to 250 degrees Fahrenheit, and a nominal micron rating of 25. It is designed to work with petroleum-based hydraulic fluids and has a height of 5.81 inches, an outside diameter of 3.74 inches, and 1-16 UNF threads. The 170HY6 filter can capture contaminants and prevent wear to help extend the life of hydraulic components.