Learn More
Grainger Externalooving Toolholder KKCR 122C
Catalog No. 502764771
Encompass_Preferred
The 170HY9 is a 3/4 HP, 115V, 1725 RPM, single phase, open drip proof motor made by Baldor Electric Company. It has a 48Y frame with a 1.15 service factor. The motor is designed for general purpose applications and features a cast iron frame and end bells along with ball bearings for smooth operation. With a continuous current rating of 7.6 amps, this motor is suitable for driving pumps, fans, conveyors, and other industrial equipment. The dimensions are 9-1/4" long by 7-3/4" wide by 8-5/8" high with a shaft diameter of 5/8". It weighs approximately 30 pounds.
Catalog No. 50-276-4771 Supplier Grainger Supplier No. 170HY9
The 170HY9 is a 3/4 HP, 115V, 1725 RPM, single phase, open drip proof motor made by Baldor Electric Company. It has a 48Y frame with a 1.15 service factor. The motor is designed for general purpose applications and features a cast iron frame and end bells along with ball bearings for smooth operation. With a continuous current rating of 7.6 amps, this motor is suitable for driving pumps, fans, conveyors, and other industrial equipment. The dimensions are 9-1/4" long by 7-3/4" wide by 8-5/8" high with a shaft diameter of 5/8". It weighs approximately 30 pounds.