Learn More
Grainger Externalooving Toolholder KKCR 203D
Catalog No. 502764773
Encompass_Preferred
The Grainger 170HZ7 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings for quiet operation. This 1/3 HP motor can provide 0.25 horsepower and is thermally protected. The Dayton brand offers a 1 year warranty on this motor. With a C-face mounting, this electric motor weighs 14 lbs and measures 3-7/8" diameter x 5-3/16" long. It operates on 60 Hz power and has a service factor of 1.0. The 170HZ7 is ideal for driving pumps, fans, and shop tools.
Catalog No. 50-276-4773 Supplier Grainger Supplier No. 170HZ7
The Grainger 170HZ7 is a Dayton 1/3 HP General Purpose Motor that runs on 115V AC power at 1725 RPM. It is a single phase, open drip proof motor with a 48Y frame and features sleeve bearings for quiet operation. This 1/3 HP motor can provide 0.25 horsepower and is thermally protected. The Dayton brand offers a 1 year warranty on this motor. With a C-face mounting, this electric motor weighs 14 lbs and measures 3-7/8" diameter x 5-3/16" long. It operates on 60 Hz power and has a service factor of 1.0. The 170HZ7 is ideal for driving pumps, fans, and shop tools.