Learn More
Grainger Externalooving Toolholder KKCR 62CF
Catalog No. 502764780
Encompass_Preferred
The 170HZ9 External Grooving Toolholder from Kennametal is a right-hand external grooving toolholder with a KKCR-62CF insert. It has a shank diameter of 0.625 inches, an overall length of 4 inches, and a maximum groove width capacity of 0.236 inches. The toolholder body is made from steel and utilizes the KKCR-62CF insert, which features a 0.062 inch thickness. This external grooving toolholder enables both axial and radial machining operations.
Catalog No. 50-276-4780 Supplier Grainger Supplier No. 170HZ9
The 170HZ9 External Grooving Toolholder from Kennametal is a right-hand external grooving toolholder with a KKCR-62CF insert. It has a shank diameter of 0.625 inches, an overall length of 4 inches, and a maximum groove width capacity of 0.236 inches. The toolholder body is made from steel and utilizes the KKCR-62CF insert, which features a 0.062 inch thickness. This external grooving toolholder enables both axial and radial machining operations.