Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Exton Qualitative Reagent, for the Qualitative Detection of Albumin in Urine, Ricca Chemical
SDP

Catalog No. p-7239355 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2950-16 500 mL
2950-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2950-16 Supplier Ricca Chemical Company Supplier No. 295016
Only null left
Add to Cart
Add to Cart

For the Qualitative Detection of Albumin in Urine

Specifications

Certifications/Compliance ISO Guide 31
CE Marker No
Clia Complexity Not Applicable
Detectable Analytes Albumin
Format Amber Glass Bottle
Sample Type Urine
Type Exton Qualitative Reagent
For Use With (Application) Qualitative detection of Albumin in Urine
Quantity 500 mL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.