Learn More
Electron Microscopy Sciences Options are available for each excitation light source.
SDP
Supplier:  Electron Microscopy Sciences FGRB2
Encompass_Preferred
The NIGHTSEA BlueStar flashlight combined with the matched barrier filter glasses is a convenient, powerful tool for exciting fluorescence in many subjects. The BlueStar uses the latest in high intensity LED technology combined with specialized optics to produce an ultra-tight, ultra-bright 10-degree beam pattern for maximum excitation of fluorescence. Comes with adjustable, cushioned wrist lanyard, rubber glare guard. Made in USA. Glasses meet ANSI Z87.1 impact standards for safety glasses.
Catalog No. NC2072524