Learn More
Research Products International Corp Extra Cap with Barb Fittings for M-Vac Jr Bottle
SDP
Supplier:  Research Products International Corp 248983
Encompass
This 2 liter capacity M-Vac Jr is the ideal size for use in most research laboratories. The M-Vac Jr aspirates a variety of liquids and supernatants such as cell culture media, reagents and other biological liquids. The M-Vac is UV resistant, unbreakable, economical and safer to use than glass collection flasks. Complete unit can be autoclaved at 120° C for 20 minutes ensuring maximum safety and withstands 700 mm Hg. The complete M-Vac Jr system includes: 2 liter polypropylene vacuum bottle, polypropylene cap with barb fittings, stainless steel stand and flexible silicone vacuum tubing.
Includes: Cap with barbs only.
Catalog No. 50-197-8388