Learn More
Fine Science Tools Extra Fine Graefe Forceps, Stainless Steel, Slightly Curved, 10 cm
SDP
Catalog No. NC1653779
Encompass
Tip Shape: Slightly Curved/ Tips: Smooth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm
This item is not returnable. View return policy
Catalog No. NC1653779 Supplier Fine Science Tools Supplier No. 1115910
Tip Shape: Slightly Curved/ Tips: Smooth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm