Learn More
Fine Science Tools Extra Fine Graefe Forceps, Stainless Steel, Curved, 10 cm
SDP
Catalog No. NC1668851
Encompass
Tip Shape: Curved/ Tips: Smooth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC1668851 Supplier Fine Science Tools Supplier No. 1116010
Tip Shape: Curved/ Tips: Smooth/ Tip Dimensions: 0.5 x 0.5mm/ Alloy / Material: Stainless Steel/ Length: 10cm