Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Extra Heavy Duty Duct Tape

Catalog No. 19072037 Shop All 3M Products
Change view
Click to view available options
Color:
Black
Silver
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-072-037 Black
19-072-095 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-072-037 Supplier 3M™ Supplier No. 7000001232
Add to Cart
Add to Cart

3M™ Extra Heavy Duty Duct Tape has a unique construction that allows for permanent and temporary indoor applications.

  • Constructed of polyethylene film laminated to cloth with a rubber adhesive
  • Resists curling and tears off the roll cleanly for easy application in MRO/construction.
  • Removable with little or no adhesive residue up to three months after application.
  • Designed for use in heavy duty MRO/construction
  • Sticks immediately and stays on reliably
  • Strong water-resistant backing resists wear and light abrasion
  • Provides a tight, moisture-resistant seal in many containment situations
  • Conforms to irregular surfaces without lifting
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Rubber
Product Type Industrial Tape
For Use With (Application) Communications, Transportation
Model 6969
Quantity 24/Cs.
Thickness (English) 10.7 mil
Dimensions (L x W) 54.8 m x 48 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.