Learn More
Kuhner Shaker Inc EXTRA HIGH UNIVERSAL CLAMP WIT
Supplier: Kuhner Shaker Inc SMCR1801CH
Extra high universal clamp with 4 positions (Kuhner EPFL only) with """"anti fly-out"""" brackets, for safer stacking of DWPs