Learn More
Kuhner Shaker Inc EXTRA HIGH UNIVERSAL PLATE CLA
Supplier:  Kuhner Shaker Inc SMCR1801A
Encompass
Extra high universal plate clamp, two DWP stacked with spacers. (Attaches to Kuhner Shaker universal tray) Spacer item # SMCR0020 not included. (4) spacers recommended.
Catalog No. 50-257-0119