Learn More
Kuhner Shaker Inc EXTRA HIGH UNIVERSAL PLATE CLA
Catalog No. 502570120
Encompass
Extra high universal plate clamp, two DWP stacked with spacers. Fits various shaker platforms. Spacer item # SMCR0020 not included. (4) spacers recommended.
Catalog No. 50-257-0120 Supplier Kuhner Shaker Inc Supplier No. SMCR1801
Extra high universal plate clamp, two DWP stacked with spacers. Fits various shaker platforms. Spacer item # SMCR0020 not included. (4) spacers recommended.