Learn More
Epredia™ Extra Hopper for the Vega 8-Hopper Laser Cassette Printer
Catalog No. 23900114 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Cassette Hopper
Catalog No. Product Type
23-900-114 Cassette Hopper
1 options
Catalog No. 23-900-114 Supplier Epredia™ Supplier No. 9001521
Add to Cart
Add to Cart
Additional cassette hopper for the Epredia™ Vega 8-Hopper Laser Cassette Printer. Holds up to 80 cassettes, enhancing capacity and workflow efficiency. Ideal for high-throughput labs needing extended, uninterrupted printing performance.
Key Features:
  • Compatible with the Epredia™ Vega 8-Hopper Laser Cassette Printer
  • Holds up to 80 cassettes for extended capacity
  • Supports quick switch between cassette colors or types
  • Reduces the need for frequent cassette reloading
  • Improves workflow efficiency in high-volume labs
  • Easy to install and integrate with existing printer setup
  • Minimizes downtime and manual intervention
Specifications
Product Type Cassette Hopper