Learn More
PATHOLOGY INNOVATIONS EXTRA LARGE CHUCK 55X36 MM
Catalog No. NC2575616
Encompass_Preferred
These stainless steel chucks are 55mm by 36mm and are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue block. Chucks can also be used warm with use of the “over-chuck freezing blocks”. These chucks were designed by professor and author of frozen section literature Dr. Stephen Peters to fit most cryostats.
Catalog No. NC2575616 Supplier PATHOLOGY INNOVATIONS Supplier No. 134
These stainless steel chucks are 55mm by 36mm and are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue block. Chucks can also be used warm with use of the “over-chuck freezing blocks”. These chucks were designed by professor and author of frozen section literature Dr. Stephen Peters to fit most cryostats.