Learn More
Chemgenes Corporation DMT-1972
SDP
Catalog No. NC1738853
Encompass
DMT-1972 is a selective 5-HT2A antagonist with an IC50 of 0.34 nM, over 10,000 nM for 5-HT2C. Useful for studying 5-HT2A's role in mental disorders, it's a crystalline solid, soluble in DMSO and DMF, and should be stored at -20°C. Molecular weight: 438.90 g/mol.
Catalog No. NC1738853 Supplier Chemgenes Corporation Supplier No. DMT1972
DMT-1972 is a selective 5-HT2A antagonist with an IC50 of 0.34 nM, over 10,000 nM for 5-HT2C. Useful for studying 5-HT2A's role in mental disorders, it's a crystalline solid, soluble in DMSO and DMF, and should be stored at -20°C. Molecular weight: 438.90 g/mol.