Description
- Preprinted with all necessary information and features a breathable design.
- Made from durable, Heavy-duty Kraft paper.
- Provides reliable handling during evidence collection and transport.
- Supports secure evidence storage.
Specifications
Specifications
| Product Type | Evidence Bag |
| Color | Brown |
| Quantity | 50/Pk. |
| Dimensions (L x W x H) | 16 x 12 x 35 in. |