Learn More
Med Vet International Extra-Long Cuttable Supracondylar Plate, 2.0mm, Left, J1076G, each
Catalog No. 5000583662
Encompass_Preferred
120mm extra-long cuttable left shaft, width 5.5mm, shaft thickness: 2mm.
Catalog No. 50-005-83662 Supplier Med Vet International Supplier No. JOR1076G
Add to Cart
Add to Cart
120mm extra-long cuttable left shaft, width 5.5mm, shaft thickness: 2mm.