Learn More
Electron Microscopy Sciences Cryostat Stainless Steel ChuckExtra Large 55 x 36 mm
SDP
Catalog No. 501900441
Encompass_Preferred
These stainless steel chucks are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue block. The stem of the chuck provides the focal point for a sharp tap resulting in an easily release of the block from the well. These chucks fit many of the major brand cryostat and can be used in most cryostats with use of an adapter. These Chuck Adapters have stem measures 20 mm L x 9 mm diameter, and fit Leica and most Microm cryostats. We also offer adapters to use these chuck for other cryostats.
Catalog No. 50-190-0441 Supplier Electron Microscopy Sciences Supplier No. 6172155
These stainless steel chucks are designed to be used cold for rapid freezing, and to maximize the gripping power required to hold the embedded tissue block. The stem of the chuck provides the focal point for a sharp tap resulting in an easily release of the block from the well. These chucks fit many of the major brand cryostat and can be used in most cryostats with use of an adapter. These Chuck Adapters have stem measures 20 mm L x 9 mm diameter, and fit Leica and most Microm cryostats. We also offer adapters to use these chuck for other cryostats.