Learn More
Brady™ Extra Secure Prolock System
Catalog No. 19103921 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Green
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-103-921 Red
19-103-923 Green
19-103-922 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-103-921 Supplier Brady™ Supplier No. 122250
Add to Cart
Add to Cart
Effectively lock out a variety of isolation points. The Brady™ Extra Secure Prolock will quickly become a staple lockout device in any facility. It tackles anything from T valve handles to electrical disconnects.
  • Designed to effectively lock out a variety of isolation points, including gate valves, T valve handles, electrical disconneccts, and more
  • Economical and commact devices are highly visible and will quickly become a staple lockout device in any facility
Specifications
Product Type Pull-tight Cable Lockout System
Color Red
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.