Learn More
Caron Products And Services Additional Stainless Steel Perforated Shelf Kit, Includes 2 shelf tracks
Catalog No. NC2464873
Encompass_Preferred
Additional Stainless Steel Perforated Shelf Kit, Includes 2 shelf tracks. Each perforated shelf is capable of supporting a uniformly distributed load of 50 lbs. For use with 25/33 CF chambers.
Catalog No. NC2464873 Supplier Caron Products And Services Supplier No. SHLF300
Additional Stainless Steel Perforated Shelf Kit, Includes 2 shelf tracks. Each perforated shelf is capable of supporting a uniformly distributed load of 50 lbs. For use with 25/33 CF chambers.