Learn More
Thomas Scientific Extra Shelf, 12.0 x 15.0", Stainless Steel
SDP
Catalog No. 501457728
Encompass
The myTemp 65 Incubator is designed to provide accurate and uniform temperature control from 0 to 60°C. The broad temperature range of the instrument expands the use of this incubator to both heating and cooling applications. A large, digital display allows for visualization of the chamber temperature and the remaining incubation time, while an internal light is included for improved visualization of the chamber. In addition, a built in, internal outlet can be used to provide power to small instruments, such as shakers, rockers, etc. With forced air technology and stainless steel inner construction, uniform heat transfer is ensured throughout the internal chamber. When set for sub-ambient temperatures, the powerful compressor is activated, quickly cooling the chamber to the desired temperature. With an exterior height of 39 inches (100cm), the incubator is designed to be floor-standing, yet is also compact enough for bench-use. (Not recommended for use on benchtops taller than 32&q
Catalog No. 50-145-7728 Supplier Thomas Scientific Supplier No. 1159P20
The myTemp 65 Incubator is designed to provide accurate and uniform temperature control from 0 to 60°C. The broad temperature range of the instrument expands the use of this incubator to both heating and cooling applications. A large, digital display allows for visualization of the chamber temperature and the remaining incubation time, while an internal light is included for improved visualization of the chamber. In addition, a built in, internal outlet can be used to provide power to small instruments, such as shakers, rockers, etc. With forced air technology and stainless steel inner construction, uniform heat transfer is ensured throughout the internal chamber. When set for sub-ambient temperatures, the powerful compressor is activated, quickly cooling the chamber to the desired temperature. With an exterior height of 39 inches (100cm), the incubator is designed to be floor-standing, yet is also compact enough for bench-use. (Not recommended for use on benchtops taller than 32&q