Learn More
Kuhner Shaker Inc EXTRA TALL TOWER FOR STACKING
Catalog No. 502570118
Encompass
Extra Tall tower for stacking of two DWP and use with EPFL tables. Spacer item # SMCR0020 not included. (4) spacers recommended.
Catalog No. 50-257-0118 Supplier Kuhner Shaker Inc Supplier No. SMCR1801C
Extra Tall tower for stacking of two DWP and use with EPFL tables. Spacer item # SMCR0020 not included. (4) spacers recommended.