Learn More
Electron Microscopy Sciences Extra Wide Lab Cart, 24.5” x 3 3/8” Footprint, 14 7/8” height
SDP
Catalog No. 501901912
Encompass_Preferred
This extra wide lab cart has a large storage area with extra wide drawers and compartments, heavy-duty casters with two locking. Top andd bottom made of HDPE with a built-in handle for easy handling. 1 large compartment with hinged acrylic door. 3 extra wide drawers to store supplies that measure 24½" (W) x 3⅜" (H) x 14⅞" (D). Cart measures measures: 36" (L) x 17" (W) x 34½" (H).
Footprint: 24.5” x 3 3/8”
Height: 14 7/8”
Catalog No. 50-190-1912 Supplier Electron Microscopy Sciences Supplier No. 6438320
This extra wide lab cart has a large storage area with extra wide drawers and compartments, heavy-duty casters with two locking. Top andd bottom made of HDPE with a built-in handle for easy handling. 1 large compartment with hinged acrylic door. 3 extra wide drawers to store supplies that measure 24½" (W) x 3⅜" (H) x 14⅞" (D). Cart measures measures: 36" (L) x 17" (W) x 34½" (H).
Footprint: 24.5” x 3 3/8”
Height: 14 7/8”